Test tube racks, half-size, Poxygrid®, SP Bel-Art
Leverandør: Bel-Art Products, a Part of SP
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
F187881301 F187881600 F187881601 F187881603 F187882000 F187882001
BELAF187881301EA 792 DKK
BELAF187881301 BELAF187881600 BELAF187881601 BELAF187881603 BELAF187882000 BELAF187882001
Test tube racks, half-size, Poxygrid®, SP Bel-Art
Stativer Tube Racks
Chemical and corrosion-resistant steel wire racks are coated with heat-cured epoxy.
  • Unaffected by organic solvents, salts, weak acids and weak alkalis
  • Will withstand a temperature range of −223 to 121 ºC; autoclavable
Excellent space-savers for use in water baths, laminar flow hoods, fume hoods or any place where space is limited.
Choice of three colours: orange for biohazard warning, green and blue for general lab use.