VWR®, Men's Coats, Cotton or Polyester/Cotton
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
113-1779EA 434 DKK
113-1779 113-1773 113-1749 113-1766 113-1767 113-1768 113-1769 113-1781 113-1772 113-1780 113-1774 113-1775 113-1776 113-1777 113-1778 113-1770 113-1746 113-1748 113-1739 113-1732 113-1734 113-1735 113-1736 113-1730 113-1738 113-1729 113-1740 113-1741 113-1742 113-1743 113-1745 113-1737 113-1747 113-1731 113-1723 113-1724 113-1725 113-1726 113-1727 113-1728 113-5988 113-5989 113-5990 113-5991 114-9341 114-9342
VWR®, Men's Coats, Cotton or Polyester/Cotton
Arbejdstøj
These reusable coats for men are either made of 100% cotton or 65% polyester/35% cotton. Both fabrics are robust, tear resistant and durable. The coats are available with a tailored or stand-up collar.
  • Concealed press stud fastening and two-step adjustable cuffs
  • Two side pockets and one breast pocket
  • Side slit for improved wearing comfort
  • Washable at 95 °C, suitable for industrial laundering
Density: 215 g/m²
Colour: White
Approved in accordance with Oeko-Tex® Standard 100.
Certificeringer: Approved in accordance with Oeko-Tex® Standard 100.