Churn sample splitters for water-sediment mixtures, SP Bel-Art
Leverandør: Bel-Art Products, a Part of SP
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
F378050004 F378050014
BELAF378050004EA 15220 DKK
BELAF378050004 BELAF378050014
Churn sample splitters for water-sediment mixtures, SP Bel-Art
Prøve opdeler
Provides an efficient method of preparing subsamples of collected water-sediment mixtures from stream flows for water quality analysis.
  • Polyethylene sample splitter consists of a thick walled cylindrical vessel with a disc churn
  • Center handle extends through the cover and a locating notch keeps it oriented in one direction
  • Small polypropylene spring-loaded spigot drains the subsamples into bottles for delivery to the water quality laboratory
  • Carrying handle makes carrying the full or empty vessel easy
  • Lid and spigot are replaceable
Water sedimentMore Product Information