Surface protection mats
Leverandør: DEUTSCH NEUMANN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
5101004 5101002 5101001 5101003 5101005
DENE5101004EA 4520 DKK
DENE5101004 DENE5101002 DENE5101001 DENE5101003 DENE5101005
Surface protection mats
Bordpapir Bordpapir
Translucent bench mats made from silicone rubber (VMQ).
  • Temperature range from −60 to +200° C
  • Short periods up to 260° C
  • Durometer 60 Shore A