Griffin beakers, PP
Leverandør: BURKLE
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
7122-0250 7122-1000 7122-3000 7122-0025 7122-5000 7122-0150 7122-0400 7122-2000 7122-0500
BURK7122-0250EA 49.9 DKK
BURK7122-0250 BURK7122-1000 BURK7122-3000 BURK7122-0025 BURK7122-5000 BURK7122-0150 BURK7122-0400 BURK7122-2000 BURK7122-0500
Griffin beakers, PP
Bægre
Laboratory beakers according to DIN 12331 and ISO 7056 (Glass) for various uses.
  • PP, highly transparent
  • Blue graduation printed on
  • Shatterproof
When bought in bulk, individual print is possible.
Certificeringer: ISO 7056