Safety helmets, uvex airwing B-WR
Leverandør: UVEX
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
9762.030 9762.130 9762.230 9762.330 9762.430 9762.530
111-8234EA 169 DKK
111-8234 111-8237 111-8253 111-8254 111-8255 111-8256
Safety helmets, uvex airwing B-WR
Hjelme Sikkerhedshjelme
Airwing B-WR safety helmet made from a polyethylene shell.
  • Three variable air vents
  • Suspension harness with rotating size adjusters for infinite width adjustment
  • Provides protection even in extremely low temperatures (−30 °C)
  • Size range: 52 to 61 cm
Certificeringer: EN 397