Dry box gloves, neoprene, SP Bel-Art
Leverandør: Bel-Art Products, a Part of SP
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
H500250544 H500250546
BELAH500250544EA 6700 DKK
BELAH500250544 BELAH500250546
Dry box gloves, neoprene, SP Bel-Art
Handsker
These neoprene gauntlets protect the user from penetration by liquid, gas or solid substances.
  • Seamless one-piece construction
  • Available as standard (non bonded neoprene) or economy version (bonded neoprene)
  • Requires clamping rings to hold gloves in place
The gloves are compatible with the Scienceware® glove box BELAH50025-0000.