Beakers, jacketed, for tempering, stainless steel
Leverandør: KGW
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2200 2201 2202 2204 2203 2205
462-1112EA 6340 DKK
462-1112 462-1114 462-1115 462-1111 KGWA2203 KGWA2205
Beakers, jacketed, for tempering, stainless steel
Bægre
Made of stainless steel.
  • Good resistance to breaking
  • Operating temperature max. 200 °C
  • Coolant ram pressure max. 0,5 bar
  • Olive welded
For tempering of liquids. The tempering medium is pumped round the double walled casing with the aid of a thermostat.