Tempering beakers
Leverandør: KGW
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1351 1352 1353 1356
462-1101EA 1530 DKK
462-1101 462-1102 462-1103 KGWA1356
Tempering beakers
Bægre
Borosilicate glass 3.3.
  • Coolant ram pressure max. 0,5 bar
  • High thermal resistance
  • Very good chemical resistance
For tempering of liquids. The tempering medium is pumped round the double walled casing with the aid of a thermostat.