Centrifugal stirring shafts, BOLA
Leverandør: Bohlender
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
C377-04 C377-08 C377-10 C377-12 C377-14 C377-16
BOHLC377-04EA 1700 DKK
BOHLC377-04 BOHLC377-08 BOHLC377-10 BOHLC377-12 BOHLC377-14 BOHLC377-16
Centrifugal stirring shafts, BOLA
Omrørere Omrøreaksler
Stirrer shafts made of PTFE-jacketed stainless steel with the stirring unit made completely of PTFE.
  • Chemical resistant
  • Temperature resistance from –200 to +250 °C
For stirring in narrow mouth vessels or in vessels with ground joint opening.