Ladies trousers, cotton, 248
Leverandør: UVEX
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
8152.901 8152.902 8152.903 8152.904 8152.905 8152.906 8152.907 8152.908 8152.909 8152.910
UVEX8152.901EA 280 DKK
UVEX8152.901 UVEX8152.902 UVEX8152.903 UVEX8152.904 UVEX8152.905 UVEX8152.906 UVEX8152.907 UVEX8152.908 UVEX8152.909 UVEX8152.910
Ladies trousers, cotton, 248
Arbejdstøj
These comfortable trousers are made from 100% cotton.
  • Elastic waistband
  • One side pocket with concealed push button
  • Machine washable until 60 °C
Weight: 210 g/m²
Certificeringer: Oeko-Tex Standard 100