Chromatographic columns with fused filter discs, VitraPOR®
Leverandør: ROBU GLASFILTER
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
31202 31101 31102 31103 31201 31203 31204 31104
ROBU31202EA 2940 DKK
ROBU31202 ROBU31101 ROBU31102 ROBU31103 ROBU31201 ROBU31203 ROBU31204 ROBU31104
Chromatographic columns with fused filter discs, VitraPOR®
Kromatografikolonner
VitraPOR® chromatography columns are made of borosilicate glass 3.3. The versatile and recyclable material makes it suitable for the most diverse fields.
  • Highly resistant to most chemicals such as solvents, acids and alkalis
  • Glass filters are divided into porosity classes ranging from 1,0 to 500 µm
Leveringsinformation: Filter tubes are available in various diameters and pore sizes.