Protease inhibitor cocktails, ProteaseArrest™
Leverandør: G Biosciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
786-108 786-329 786-437 786-711 786-712
733-1996EA 2730 DKK
733-1996 786-329 786-437 786-711 GENO786-712
Protease inhibitor cocktails, ProteaseArrest™
Celledyrkningssupplementer og additiver
The ProteaseArrest™ family of protease inhibitor cocktails are supplied as convenient, ready-to-use, liquid format cocktails that offer immediate >95% inhibition of harmful proteases. The 100X concentrated solution outperforms other similar protease inhibitor cocktails (including tablet formats) by offering instant inhibition, allowing delivery of optimised concentrations, demonstrating higher inhibition levels, and providing a greater ease of use. Also available as single use OneQuant™ ProteaseArrest™ aliquots.
  • Ready-to-use, broad spectrum, 100X concentrated protease inhibitor cocktail
  • High inhibition levels: 1X ProteaseArrest™ inhibits >95% of protease activities
  • Outperforms tablet cocktails
  • Excellent inhibition of protease activity in preparations of mammalian, bacteria, plant, yeast and fungal lysates
  • Protection of proteins in such applications as electrophoresis, purification, storage, assays and more
  • Suitable for small volume, analytical sample applications
Bestil nu
Specifikationer
Environmentally Preferable
Species
Kilde