4-tert-Butylcalix(4)aren ≥99%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
B21515.06
B21515.14
B21515.06EA
691
DKK
B21515.06
B21515.14
4-tert-Butylcalix(4)aren ≥99%
4-tert-Butylcalix(4)aren
Purity: 99%
CAS No.: 60705-62-6
Molecular Formula: C44H56O4
Formula Weight: 648.93
Melting Point: 339-344°
Beilstein Reference: 2033965
Formel:
C₄₄H₅₆O₄ Smeltepunkt: 339…344 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00066280 CAS-nummer: 60705-62-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...