(Iodmethyl)cyclopentan ≥98% stabilised

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel


Stabilization: Stabilised with copper

B25391.14 B25391.06
B25391.14EA 7300 DKK
B25391.14 B25391.06
(Iodmethyl)cyclopentan ≥98% stabilised
(Iodomethyl)cyclopentane

Stabilized with copper


Purity: 98+% CAS No.: 27935-87-1 Molecular Formula: C6H11I Formula Weight: 210.06 Flash Point: 92°(198°F) Refractive Index: 1.542 Boiling Point: 78°/17mm MDL No.: MFCD03453190

Formel: C₆H₁₁I
Kogepunkt: 78 °C (17 mmHg)
Massefylde: 1,614
Flammepunkt: 92 °C (198 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD03453190
CAS-nummer: 27935-87-1

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR