Dimethylarsinsyre natriumsalt trihydrat

Leverandør: Serva
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Dimethylarsinic acid sodium salt trihydrate, Sodium cacodylate trihydrate, Dimethylarsonic acid sodium salt trihydrate, Sodium dimethylarsinate trihydrate

15540.02 15540.03
SERA15540.02EA 738.95 DKK
SERA15540.02 SERA15540.03
Dimethylarsinsyre natriumsalt trihydrat
Dimethylarsinsyre natriumsalt trihydrat
Formel: (CH₃)₂As(O)ONa·3H₂O
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00149079
CAS-nummer: 6131-99-3
EINECS: 204-708-2
UN nummer: 1688
ADR: 6.1,II

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR