1-Methyl-1-phenylhydrazin ≥97%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
B22440.06
B22440.14
B22440.22
B22440.06EA
1310
DKK
B22440.06
B22440.14
B22440.22
1-Methyl-1-phenylhydrazin ≥97%
1-Methyl-1-phenylhydrazine
Purity: 97%
CAS No.: 618-40-6
Molecular Formula: C7H10N2
Formula Weight: 122.17
Flash Point: 92°(198°F)
Refractive Index: 1.5830
Boiling Point: 54-55°/0.3mm
MDL No.: MFCD00007627
Formel:
C₇H₁₀N₂ Kogepunkt: 54…55 °C (0,3 mmHg) Massefylde: 1,038 Flammepunkt: 92 °C (198 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00007627 CAS-nummer: 618-40-6 EINECS: 210-547-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...