(2-Hydroxyethyl)hydrazin
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
β-Hydroxyethylhydrazine
43793.09
43793.30
43793.18
43793.09EA
473
DKK
43793.09
43793.30
43793.18
(2-Hydroxyethyl)hydrazin
(2-Hydroxyethyl)hydrazine
CAS No.: 109-84-2
Molecular Formula: HOCH2CH2NHNH2
Formula Weight: 76.10
Flash Point: 73°(163°F)
Physical Form: Liquid
Density: 1.121
Refractive Index: 1.4930
Melting Point: -70°
Boiling Point: 218-220°
UN No.: 2810
MDL No.: MFCD00007623
Formel:
C₂H₈N₂O Kogepunkt: 218…220 °C Smeltepunkt: –70 °C Massefylde: 1,121 Flammepunkt: 73 °C (163 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00007623 CAS-nummer: 109-84-2 EINECS: 203-711-6 UN nummer: 2810 ADR: 6.1,II |