Triethylenetetramine mixture of isomers ≥95% for synthesis, Sigma-Aldrich®

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: N,N'-Bis(2-aminoethyl)-1,2-ethanediamine, Trientine, DEH 24, EPH 925

M-Clarity™ quality level = MQ200

8.14392.0100 8.14392.0500
8.14392.0100EA 191 DKK
8.14392.0100 8.14392.0500
Triethylenetetramine mixture of isomers ≥95% for synthesis, Sigma-Aldrich®
Triethylenetetramine
Formel: C₆H₁₈N₄
Molekylvægt: 146.24 g/mol
MDL Number: MFCD00008169
CAS-nummer: 112-24-3
EINECS: 203-950-6
UN nummer: 2259
ADR: 8 II

Bestil nu

Specifikationer Testresultater

Solubility (20 °C) soluble
Melting Point 12 °C
Molar Mass 146.23 g/mol
Boiling Point 270 - 300 °C (1013 hPa)
Vapor Pressure  - 0.01 hPa (20 °C)
Flash Point 129 °C
Refractive Index 1.4971 (20 °C, 589 nm)
Explosion Limit 0.7 - 7.2 %(V)
Density 0.98 g/cm3 (20 °C)
pH 10 - 11 (10 g/l, H2O, 20 °C)
Ignition Point 335 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR