Tantalstandard, 10g/L Ta i 2% flussyre (from TaCl₅) Specpure® enkeltelement standard til ICP
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
14419.AD
14419.AE
14419.AP
14419.ADEA
1720
DKK
14419.AD
14419.AE
14419.AP
Tantalstandard, 10g/L Ta i 2% flussyre (from TaCl₅) Specpure® enkeltelement standard til ICP
Standarder
Kationiske standarder
Tantalum Standards
Molecular Formula: TaCl₅ in 2% HF
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011252
Certificeringer: Traceable to NIST