Methylcyclopentadiene dimer tech. 90% stabilised, Technical Grade
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Stabilization:
stabiliseret med 4-tert-Butylpyrocatechol 200 ppm
A17596.0J
A17596.AP
A17596.AU
A17596.0JEA
3670
DKK
A17596.0J
A17596.AP
A17596.AU
Methylcyclopentadiene dimer tech. 90% stabilised, Technical Grade
Methylcyclopentadiene dimer
Purity: 90%
CAS No.: 26472-00-4
Molecular Formula: C12H16
Formula Weight: 160.26
Flash Point: 26°(79°F)
Refractive Index: 1.4980
Melting Point: ≈-50°
Boiling Point: 198-201°
UN No.: 3295
MDL No.: MFCD00082297
Formel:
C₁₂H₁₆ Kogepunkt: 198…201 °C Smeltepunkt: <–50 °C Massefylde: 0,940 Flammepunkt: 26 °C (79 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00082297 CAS-nummer: 26472-00-4 EINECS: 247-724-5 UN nummer: 3295 ADR: 3,III |