1-(Methylamino)anthraquinon 98%, Sigma-Aldrich®

Leverandør: SIGMA ALDRICH MICROSCOPY
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 1-(Methylamino)anthracene-9,10-dione

M-Clarity™ quality level = MQ100

M28200-5G M28200-100G M28200-500G
M28200-5GEA 436 DKK
M28200-5G M28200-100G M28200-500G
1-(Methylamino)anthraquinon 98%, Sigma-Aldrich®
1-(Methylamino)anthraquinon
Formel: C₁₅H₁₁NO₂
Molekylvægt: 237.26 g/mol
Smeltepunkt: 170...172 °C (lit.)
Opbevaringstemperatur: Room Temperature
MDL Number: MFCD00001197
CAS-nummer: 82-38-2
EINECS: 201-417-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6faee21e54c59d5557eae3a273a42330a2b3a50a&vid=6e88f2082a2422525584fa56e8c3c336de019c58&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance (colour) Faint red to very dark red
Appearance (form) Powder or crystals
Purity (HPLC area %) ≥ 97.5 %
Carbon content 74.0 - 77.8 %
Nitrogen content 5.6 - 6.1 %
Infrared spectrum Conforms to structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR