Colour standard solutions, Pt-Co (Hazen)
Leverandør: REAGECON
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
HAZ250 HAZ10 HAZ25 HAZ50 HAZ0 HAZ100 HAZ40 HAZ500 HAZ5 HAZ5005
634-0500EA 3270 DKK
634-0500 REGEHAZ10 REGEHAZ25 REGEHAZ50 REGEHAZ0 REGEHAZ100 REGEHAZ40 REGEHAZ500 REGEHAZ5 REGEHAZ5005
Colour standard solutions, Pt-Co (Hazen)
Standarder Farve Referenser og Spektroskopi Standarder
The products can be used to calibrate, control, qualify and validate colour measurement instruments.
  • Ready to use
  • Prepared gravimetrically on a weight/weight basis.
  • Consistency of product - independent, traceable, certified