Cyclopentadecan ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

A19416.03 A19416.06
A19416.03EA 416 DKK
A19416.03 A19416.06
Cyclopentadecan ≥98%
Cyclopentadecane

Purity: 98% CAS No.: 295-48-7 Molecular Formula: C15H30 Formula Weight: 210.40 MDL No.: MFCD00039424

Formel: C₁₅H₃₀
Molekylvægt: 210.4 g/mol
Massefylde: 0.805
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00039424
CAS-nummer: 295-48-7
EINECS: 206-040-7

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR