5-Chlor-2-fluorbenzaldehyd ≥97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

H26187.03 H26187.06 H26187.14
H26187.03EA 633 DKK
H26187.03 H26187.06 H26187.14
5-Chlor-2-fluorbenzaldehyd ≥97%
5-Chlor-2-fluorbenzaldehyd

Purity: 97% CAS No.: 96515-79-6 Molecular Formula: C7H4ClFO Formula Weight: 158.56 Refractive Index: 1.5410 Melting Point: 29-32° MDL No.: MFCD03094518

Formel: C₇H₄ClFO
Smeltepunkt: 29…32 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD03094518
CAS-nummer: 96515-79-6
EINECS: None

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR