Lanthanstandard, 10g/L La i 5% salpetersyre (from La₂O₃) Specpure® enkeltelement standard til ICP
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
14429.AP
14429.AD
14429.AE
14429.APEA
4180
DKK
14429.AP
14429.AD
14429.AE
Lanthanstandard, 10g/L La i 5% salpetersyre (from La₂O₃) Specpure® enkeltelement standard til ICP
Standarder
Kationiske standarder
Lanthanum Standards
Molecular Formula: La₂O₃ in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011066
Certificeringer: Traceable to NIST