Palladiumstandard, 10g/L Pd i 20% saltsyre (from Pd) Specpure® enkeltelement standard til ICP
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
14432.AD
14432.AE
14432.AP
14432.ADEA
4000
DKK
14432.AD
14432.AE
14432.AP
Palladiumstandard, 10g/L Pd i 20% saltsyre (from Pd) Specpure® enkeltelement standard til ICP
Standarder
Kationiske standarder
Palladium Standards
Molecular Formula: Pd in 20% HCl
Physical Form: Liquid
Odour: Characteristic
UN No.: 1789
MDL No.: MFCD03613603
Certificeringer: Traceable to NIST