Nikkelstandard, 1g/L Ni i 5% salpetersyre (from Ni) Specpure® standard til AAS
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
88082.AE
88082.AP
88082.AEEA
517
DKK
88082.AE
88082.AP
Nikkelstandard, 1g/L Ni i 5% salpetersyre (from Ni) Specpure® standard til AAS
Standarder
Kationiske standarder
Nickel Standards
Molecular Formula: Ni in 5% HNO₃
Physical Form: Liquid
Appearance: Colourless
UN No.: 3264
MDL No.: MFCD00011137
Certificeringer: Traceable to NIST