Manganstandard, 1g/L Mn i 5% salpetersyre (from Mn) Specpure® standard til AAS
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
88078.AE
88078.AP
88078.AEEA
512
DKK
88078.AE
88078.AP
Manganstandard, 1g/L Mn i 5% salpetersyre (from Mn) Specpure® standard til AAS
Standarder
Kationiske standarder
Manganese Standards
Molecular Formula: Mn in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011111
Certificeringer: Traceable to NIST