Iod(I)chlorid 1 M in dichloromethane
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Farlig
Synonymer: Iodochloride
H31938.AE H31938.AP
H31938.AEEA 797 DKK
H31938.AE H31938.AP
Iod(I)chlorid 1 M in dichloromethane
Iodine monochloride
CAS No.: 7790-99-0 Molecular Formula: ClI Formula Weight: 162.36 UN No.: 2922 MDL No.: MFCD00011354
Formel: ClI
Molekylvægt: 162.36 g/mol
Kogepunkt: 97.3
Smeltepunkt: 26
Massefylde: 1.420
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011354
CAS-nummer: 7790-99-0
EINECS: 232-236-7
UN nummer: 2922
ADR: 8 III
Merck Index: 13,05039