4-Propoxyanilin ≥97%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
H55096.03
H55096.06
H55096.03EA
545
DKK
H55096.03
H55096.06
4-Propoxyanilin ≥97%
4-Propoxyanilin
Formel:
C₉H₁₃NO Flammepunkt: 110 °C (230 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00043621 CAS-nummer: 4469-80-1 EINECS: 224-740-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...