trans-4-Methyl-2-penten ≥95.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: (E)-4-Methyl-2-pentene

M0394-25ML M0394-5ML
TCIAM0394-25MLEA 3640 DKK
TCIAM0394-25ML TCIAM0394-5ML
trans-4-Methyl-2-penten ≥95.0% (by GC)
trans-4-Methyl-2-pentene
Formel: C₆H₁₂
Molekylvægt: 84.16 g/mol
Kogepunkt: 58°C
Smeltepunkt: -140.8
Flammepunkt: –27°C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00065138
CAS-nummer: 674-76-0
EINECS: 211-616-6
UN nummer: 2288
ADR: 3 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=43e61046075f4d1e4e92c1187bcca1e1841b3e0e&vid=d73731c267fe74b9c0b95b8328e7a5f801032679&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR