4-Fluor-N-methylanilin ≥97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: N-(4-Fluorophenyl)-N-methylamine

H55105.03 H55105.06
H55105.03EA 480 DKK
H55105.03 H55105.06
4-Fluor-N-methylanilin ≥97%
4-Fluor-N-methylanilin

Purity: 97% CAS No.: 459-59-6 Molecular Formula: C7FH8N Formula Weight: 125.15 Flash Point: 76°(169°F) Refractive Index: 1.5320 Boiling Point: 79°/11mm UN No.: 2810 MDL No.: MFCD00017942

Formel: C₇H₈FN
Molekylvægt: 125.15 g/mol
Kogepunkt: 79 °C (11 mmHg)
Massefylde: 1.040
Flammepunkt: 76 °C (169 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00017942
CAS-nummer: 459-59-6
EINECS: 207-294-1
UN nummer: 2810
ADR: 6.1 III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR