Oxacyclohexan for synthesis, Sigma-Aldrich®

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig


M-Clarity™ quality level = MQ200

8.43950.0250 8.43950.1000
8.43950.0250EA 848 DKK
8.43950.0250 8.43950.1000
Oxacyclohexan for synthesis, Sigma-Aldrich®
Tetrahydropyran
Formel: C₅H₁₀O
Kogepunkt: 88 °C (1013 hPa)
Massefylde: 0,881 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00006585
CAS-nummer: 142-68-7
EINECS: 205-552-8
UN nummer: 3271
ADR: 3,II
Merck Index: 12,09356

Bestil nu

Specifikationer Testresultater

Solubility 80.2 g/l (25°C)
Melting Point -49.2 °C
Boiling Point 88 °C (1013 hPa)
Flash Point -16 °C
Density 0.88 g/cm3 (25 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR