Flasker, bred hals, firkantede, med skruelåg
Leverandør: LAMAPLAST
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
614 613 612 611 610 609 608
215-9004EA 72.8 DKK
215-9004 215-9003 215-9002 215-9001 215-9000 215-0916 215-0915
Flasker, bred hals, firkantede, med skruelåg
Flasker Plastflasker
HDPE, translucent.
  • Good chemical resistance
  • Heat resistant up to 80 °C
  • Leakproof, graduated
Double-closure cap, shoulder and perforated cap allow the installation of seals and labels to ensure the integrity of the contents. Can be used anywhere in the laboratory and for sampling. Wide mouth for rapid filling and dispensing of powders, pastes and liquids.