Mortars without pestles, SP Bel-Art
Leverandør: Bel-Art Products, a Part of SP
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
H372600000 H372600001
BELAH372600000EA 7530 DKK
BELAH372600000 BELAH372600001
Mortars without pestles, SP Bel-Art
Mortere og pistiller Mortere
Liquid nitrogen cooled mortars without pestles, for grinding frozen samples.
  • Insulated handle is removable to save space; 10,2cm long
  • Three suction cup feet stabilise the unit on the workbench
  • Recommended size pestle is any size appropriate for a 275ml mortar (not included)
Grind frozen samples without liquid nitrogen hazards. This 304 stainless steel mortar is suspended by three brackets above a stainless steel bowl which holds liquid nitrogen. The bowl is nested in a heavily insulated blue polyethylene housing.