Reusable earplugs, uvex whisper+
Leverandør: UVEX
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2111.212 2111.238
111-1572EA 945 DKK
111-1572 UVEX2111.238
Reusable earplugs, uvex whisper+
Ørepropper Genanvendelige ørepropper
These reusable plugs feature a smooth, dirt-repellent surface which can be cleaned using soap and water, maintaining the wearer's comfort and safety.
  • Pre-shaped: Allows easy insertion
  • Soft material: Very comfortable to wear, even during extended use
  • Unique design: combination of straight and curved ridges reduce undesired transmission and enhance comfort
  • Smaller diameter of stems ensures optimum wearer comfort
  • Flexible wearing style with removable cord
  • Hygienic storage in an individual carrying case
Suitable for use in dirty working environments.
SNR: 27 dB
Certificeringer: EN 352-2
Advarsel: * One size fits all.