Centrifuge tubes, conical, Nunc™
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
339650 339651 339653 339652 376814
525-0492EA 2530 DKK
525-0492 525-0493 525-0495 525-0494 525-0915
Centrifuge tubes, conical, Nunc™
Rør Centrifugerør
PP, conical bottom, graduated, sterile, autoclavable with HDPE screw cap.
  • Leakproof to protect samples
  • USP Class VI, non pyrogenic, non cytotoxic and RNase-/DNase-free
  • Recyclable plastic rack reduces waste and minimises risk of contamination and particles
It provides open, easy access for pipetting, decanting, and removal of the pellet following centrifugation.
Certificeringer: USP Class VI.
Emballering: Tubes are packaged 40 per case.