PerfeCTa® QPCR mixes, SYBR® green super mixes and fast mixes
Leverandør: Quantabio
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
95071-012 95071-05K 95072-012 95072-05K 95072-250 95073-012 95073-05K 95073-250 95074-012 95074-05K 95074-250 95054-02K 95054-500 95055-02K
733-1378EA 7450 DKK
733-1378 733-1379 733-1382 733-1383 733-1381 733-1386 733-1387 733-1385 733-1390 733-1391 733-1389 733-1247 733-1246 733-1251
PerfeCTa® QPCR mixes, SYBR® green super mixes and fast mixes
Nukleinsyrereagenser qPCR/RT-qPCR Enzymes and Kits
PerfeCTa® SYBR® Green SuperMixes and FastMixes™ enable efficient, sensitive and precise quantitative PCR with proprietary buffers and SYBR® Green stabilisers that maximise fluorescent signal, PCR efficiency, and reduce primer dimers. These SuperMixes and FastMixes™ have been optimised for all Real-Time PCR instrument platforms, including those requiring normalisation with ROX reference dye or fluorescein.
  • FastMixes™ offer shorter run times enabling more experiments per day
  • Broad dynamic range ensures data from precious samples is more reliably attained
  • Superior antibody-mediated hot start results in higher specificity leading to more accurate quantification
  • Fast Cycling using existing primer sets means no re-optimisation and increased productivity
PerfeCTa® SYBR® Green SuperMixes are 2X concentrated, ready to use reaction cocktails containing all components for qPCR, except primers and template. A key component of these SuperMixes is AccuStart™ Taq DNA polymerase, which enables specific and efficient primer extension with the convenience of room temperature reaction assembly. AccuStart™ Taq DNA polymerase contains monoclonal antibodies that bind to the polymerase and keep it inactive prior to the initial PCR denaturation step. Upon heat activation the antibodies denature irreversibly, releasing fully active and unmodified Taq DNA polymerase.
PerfeCTa® SYBR® Green FastMixes™ are 2X concentrated, ready to use reaction mixes delivering maximum PCR efficiency, sensitivity, specificity and robust fluorescent signal using either fast or conventional cycling protocols. Rapid cycling is achieved by instant activation of AccuStart™ Taq DNA polymerase coupled with rapid polymerisation kinetics. High performance qPCR can be achieved in as little as 33 minutes.