WHEATON® Container, Wide Mouth, with Cap
Leverandør: DWK Life Sciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
W209904 W209905 W209900 W209906 W209902 W209903
WHEAW209904EA 1500 DKK
WHEAW209904 WHEAW209905 WHEAW209900 WHEAW209906 WHEAW209902 WHEAW209903
WHEATON® Container, Wide Mouth, with Cap
Prøvecontainere Allround kasser
Container made of PP, with linerless cap.
  • Wide mouth design is ideal for large solid samples
  • Durable with good chemical resistance
  • White screw cap pre-attached to maintain cleanliness
  • Sterilised using ethylene oxide