Readycult®
Leverandør: Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1.01295.0001 1.01298.0001 1.01299.0001
1.01295.0001EA 889 DKK
1.01295.0001 1.01298.0001 1.01299.0001
Readycult®
Mikrobiologiske medier Klargjort medium
Readycult® Coliform and Readycult® Enterococci make it easy to detect coliforms, E. coli or faecal streptococci in water. You only need a sterile container and a UV lamp (Cat. No. 1.13203.0001).
  • Easy handling: Readycult® is ready to use in a blister pack
  • Quick results: Add Readycult® to the water sample straight from the blister pack, mix, incubate for 24 ±1 hour, read result, finished!
  • Safe and easy interpretation: Readycult® reliably detects even damaged organisms. Blue-green colour change shows the presence of Total coliforms, E. coli or Enterococci (Cat. No. 1.01299); fluorescence + indole reaction shows the presence of E. coli. * for 50 ml water; ** for 100 ml water