Dip slides
Leverandør: LIOFILCHEM
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
525622 525282 525302 525312 525272
LIOF525622EA 432 DKK
LIOF525622 LIOF525282 LIOF525302 LIOF525312 LIOF525272
Dip slides
Mikrobiologiske medier Klargjort medium
Two-sided slide with agar-agar centre. Operating instructions: 1. Unscrew the cap containing the agar-agar slide. Hold the bottle in the other hand and lightly apply pressure to the sampling area using the agar-agar slide; or 2. Apply directly to the middle of the swab brush if you have completed a swab sample; or 3. Immerse the agar-agar slide in the liquid to analyse. 4. Insert the slide into the bottle and screw back on. Place in the oven at 37 °C for 24/48 hours. To detect yeasts, increase the incubation by 24/48 hours at 25/30 °C.
  • Box of 10 slides with separate labels.
  • Use-by date: six months after manufacture
  • Storage: ambient temperature (max. 25 °C)