VWR® Culture Media, Ready to Use, in Bags
Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
AX031108EA 0 DKK
AX031108 AX031106 AX031107 AX031109 AX031110 AX031119 800179ZB 800343VB 801121ZB 850179ZB 850343VB 853120ZB 870179ZB 870343VB 851121ZB 803120ZB 852760TA 872760TA 870343GB 800179ZT 870179ZBMP 870343VBMP 850343VBMP 800179ZBMP 800343VBMP 850179ZBMP 801121ZE 806354SA 856354SA 850179ZBB 871051ZB 871121ZB 850343VBB
VWR® Culture Media, Ready to Use, in Bags
Mikrobiologiske medier Klargjort medium
Mikrobiologiske medier i poser er ideelle til store prøvevolumener.
  • Space saving packaging
  • No risk associated with broken glass bottles
  • Media can be easily and conveniently dispensed using a gravimetric diluter or peristaltic pump