VWR®, MediaBag Dehydreret kulturmedie i poser
Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
7201.03600EA 4520 DKK
7201.03600 7201.04500 7201.06750 7203.00372 7301.00225 7501.00225 7301.00900 7301.01125 7801.01350 7601.04000 7601.03375 7200.00001 7511.00225 7201.09000 7501.00450 7504.02250 7504.04500 7291.04500 7291.06750 7591.00225 7205.04500 7801.60000 7301.02250 7601.04500 7601.03000 7503.03375 7304.00225 7301.00090 7303.00225 7812.05000 7503.00225 7504.00450 7504.00045 7891.60000 7583.0005 7812.06000 7812.11100 7301.02700 7203.03375 7691.03375 7201.02700 7201.03375 7301.00450 7812.00600 7812.09000 7812.00500 710-2051 710-2052 710-2056 710-2057 710-2058 710-2059 710-2060 710-2061
VWR®, MediaBag Dehydreret kulturmedie i poser
Mikrobiologiske medier Klargjort medium
Hver pose indeholder en afvejet mængde granuleret medie emballeret i to lag poser og bestrålet. Tidsbesparende opformering - tilsæt sterilt vand og prøvemateriale
  • Pladsbesparende, ingen flasker, som bruger hyldeplads
  • Lang holdbarhed
  • Ingen autoklavering og optimal opløselighed
  • Støvfrit
The following media are available for the bag system:
Buffered peptone water (BPW), in compliance with ISO 6579-1:2017, is a non selective pre-enrichment broth to increase recovery of bacteria, particularly pathogenic Enterobacteriaceae from foodstuffs
Tryptic soy broth (TSB) used mainly for Media Fill test
Fraser broth base used for the selective enrichment of Listeria spp.
Lactose broth used as a preliminary test for coliform bacteria detection
MacConkey broth for the detection and enumeration of coliforms
Non selective pre-enrichment broth to increase recovery of bacteria, particularly pathogenic Enterobacteriaceae from foodstuffs. The culture medium complies with ISO 6579:2002.
The lab-blender bag contains 5,75 g of BPW to which 225 ml of sterile water and 25 g (or 25 ml) of sample material is added.
More media are available on request.
The following media are approved for the bag system:
Certificeringer: Sammensætning i overensstemmelse med standardmetoder, såsom ISO, AFNOR, NMKL etc.
Emballering: Specielle formuleringer og emballageformater fås på anmodning.