Certified Reference Materials for environmental analysis
Leverandør: VKI WATER QUALITY INSTITUTE
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
QCDWB QCRW1-10 QCORGSLUD100 QCORGSLUD5X100 QCWW6-50 QCRW2-50 QCMETALLL1 QCMETALLL2 QCMETALHL1 QCMETALHL2 QCWW4-50 QCMETALLL3 QCWW2.2-50 QCWW2.1-50 QCDW4-50 QCDW4-10 QCWW1B-10 QCWW1B-50 QCWW3-10 QCWW3-50 QCWW4A-50 QCWW5-50 QCRW2-10 QCMETALCOMBI QCWW2.1-10 QCWW2.2-10 QCWW4-10 QCWW4A-10 QCWW5-10 QCRW1-50 QCWW6-10 QCLOAMSOILB200 QCSW3.1B QCSW3.2B
QCDWBEA 13960 DKK
QCDWB QCRW1-10 QCORGSLUD100 QCORGSLUD5X100 QCWW6-50 QCRW2-50 QCMETALLL1 QCMETALLL2 QCMETALHL1 QCMETALHL2 QCWW4-50 QCMETALLL3 QCWW2.2-50 QCWW2.1-50 QCDW4-50 QCDW4-10 QCWW1B-10 QCWW1B-50 QCWW3-10 QCWW3-50 QCWW4A-50 QCWW5-50 QCRW2-10 QCMETALCOMBI QCWW2.1-10 QCWW2.2-10 QCWW4-10 QCWW4A-10 QCWW5-10 QCRW1-50 QCWW6-10 QCLOAMSOILB200 QCSW3.1B QCSW3.2B
Certified Reference Materials for environmental analysis
Standarder Miljø analyse standarder
Certified Reference Materials (CRMs), produced by VKI, are designed to provide appropriate homogenous stable reference materials for quality control of environmental analyses. Certification is carried out in accordance with ISO Guide 30, 31, 33, 34, 35, ISO 5725 and ISO/REMCO N 37. All CRMs are traceable to a National Standard, or verified by a group of internationally recognised laboratories, and come with a Certificate of Analysis which states the certified value and the uncertainty at a stated level of confidence.
  • Liquid references supplied in packs of glass ampoules ready for easy dilution