Corning® Endothelial Cell Growth Supplement, Corning
Leverandør: Corning
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
356006 354006
734-2391EA 3850 DKK
734-2391 734-1306
Corning® Endothelial Cell Growth Supplement, Corning
Celledyrkningssupplementer og additiver
Vækstsupplement til endothelialceller (ECGS) er et bredt anvendt supplement til dyrkningen af en række celler, især endothelialceller. ECGS indeholder forskellige vækstfaktorer (for eksempel sur FGF eller ECGF-a).
ECGS contains various growth factors (e.g., acidic FGF or ECCF-a).
Tested for ability to promote proliferation of Fetal Bovine Heart Endothelial Cells (FBHEC) in medium containing 10% newborn calf serum. Filtered (0.2 µm membrane) and tested and found negative for the presence of bacteria, fungi, and mycoplasma.