Caps for culture media bottles
Leverandør: DWK Life Sciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
290101108 290132407 290100909 290122406
391-0911EA 58.5 DKK
391-0911 214-1169 391-0910 391-0950
Caps for culture media bottles
Låg Flaskelåg
Stainless steel or Aluminium anodised.
  • Very good temperature resistance
  • Good chemical resistance
For cell culture tubes with straight rim or culture media bottles with straight neck.