Terasaki dispensers
Leverandør: HAMILTON BONADUZ
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
201900 201910 201920 201930 201940
HAMI201900EA 8190 DKK
HAMI201900 HAMI201910 HAMI201920 HAMI201930 HAMI201940
Terasaki dispensers
Sprøjtedispenseringssystemer
This multi-syringe dispenser consists of six identical 1700 RN series Gastight® syringes. An easy to use mechanism releases step by step movement of the plungers. Each movement dispenses 1/50th of the syringe volume.
  • Accurately dispense six samples simultaneously up to 50 times with the press of a lever
  • Designed for rapid filling of 60 well HLA trays with 6,35 mm well spacing
  • Removable needles (RN)
  • Plunger assemblies and glass barrels are replaceable
Typical applications includes filling of Terasaki plates, serology for transplantations, cell analysis and drop sampling.