Cover glasses, #1
Leverandør: MARIENFELD
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
0101112 0101192 0101172 0101222 0101242
631-0715EA 503 DKK
631-0715 631-0613 631-0691 631-0662 631-0664
Cover glasses, #1
Dækglas
These cover glasses are suitable for fluorescence microscopy and in vitro diagnostic (IVD) applications.
  • Long storage, non corroding
  • Made of clear borosilicate D 263™ M glass, Hydrolytic Class I
  • Do not stick together
  • Excellent flat, smooth surfaces
Certificeringer: IVD 98/79/EC.
Emballering: In plastic boxes with hinged lid – 1000 pieces per box.