Diagnostic bottles, narrow neck, round, PET, with thread, WHEATON®
Leverandør: DWK Life Sciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
W013973 W013974 W013975
WHEAW013973EA 0 DKK
WHEAW013973 WHEAW013974 WHEAW013975
Diagnostic bottles, narrow neck, round, PET, with thread, WHEATON®
Flasker Plastflasker
PET, clear or amber.
  • Provided non-sterile
  • Can be sterilized using EtO or gamma radiation
  • Lined Poylpropylene screw closures
Ideal for storage and shipment of liquid reagents, buffer solutions,
Bestillingsoplysninger: Closures sold separately.
Emballering: Packaged in shrink wrap modules